Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tubastatin A hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A107990-10MG A107990-1MG A107990-50MG A107990-5MG A107990-100MG
76982-538EA 28 USD InStock
76982-538 76982-540 76982-542 76982-544 77682-990
Tubastatin A hydrochloride 98%, Ambeed.Inc
Tubastatin A hydrochloride
Precaución: For research use only.


Formula: C₂₀H₂₁N₃O₂·HCl
MDL Number: MFCD20488052
CAS Number: 1310693-92-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty