Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(3-Cyanophenyl)nicotinic acid 95%

Supplier: AMBEED, INC
Warning

A108852-1G A108852-250MG
76982-604EA 243 USD InStock
76982-604 76982-606
6-(3-Cyanophenyl)nicotinic acid 95%
6-(3-Cyanophenyl)nicotinic acid
Precaución: for Research Use Only


Formula: C₁₃H₈N₂O₂
MW: 224.22 g/mol
MDL Number: MFCD06410410
CAS Number: 887975-97-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty