Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-3-(3-benzothienyl)-D-alanine 95%

Supplier: AMBEED, INC
Warning

A109566-1G A109566-25G A109566-5G
76995-126EA 39 USD InStock
76995-126 76995-128 76995-130
Boc-3-(3-benzothienyl)-D-alanine 95%
Boc-3-(3-benzothienyl)-D-alanine
Precaución: for Research Use Only


Formula: C₁₆H₁₉NO₄S
MDL Number: MFCD00079679
CAS Number: 111082-76-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty