Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Hydroxyphenyl)cyclohexanone 95%

Supplier: AMBEED, INC
Warning

A109875-10G A109875-1G A109875-25G A109875-5G A109875-100G
76995-148EA 19 USD InStock
76995-148 76995-150 76995-152 76995-154 77670-558
4-(4-Hydroxyphenyl)cyclohexanone 95%
4-(4-Hydroxyphenyl)cyclohexanone
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₂
MDL Number: MFCD00210693
CAS Number: 105640-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty