Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-(4-Phenoxyphenyl)-1-(piperidin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine 98%

Supplier: AMBEED, INC
Warning

A110162-10MG A110162-1G A110162-25G A110162-2MG A110162-50MG A110162-5G A110162-5MG
77008-868EA 10 USD InStock
77008-868 77008-870 77008-872 77008-874 77008-876 77008-878 77008-880 77008-882
(R)-3-(4-Phenoxyphenyl)-1-(piperidin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine 98%
(R)-3-(4-Phenoxyphenyl)-1-(piperidin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Precaución: for Research Use Only


Formula: C₂₂H₂₂N₆O
MW: 386.45 g/mol
MDL Number: MFCD20482137
CAS Number: 1022150-12-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty