Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N(α)-Fmoc-D-tryptophan 97%

Supplier: AMBEED, INC
Warning

A110273-10G A110273-25G A110273-5G A110273-500G
76995-166EA 9 USD InStock
76995-166 76995-168 76995-170 77670-564
N(α)-Fmoc-D-tryptophan 97%
N(α)-Fmoc-D-tryptophan
Precaución: for Research Use Only


Formula: C₂₆H₂₂N₂O₄
MW: 426.46 g/mol
MDL Number: MFCD00062954
CAS Number: 86123-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty