Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-N,N-dimethylpyridin-3-amine 95%

Supplier: AMBEED, INC
Warning

A110973-100MG A110973-1G A110973-250MG A110973-25G A110973-5G
77008-912EA 12 USD InStock
77008-912 77008-914 77008-916 77008-918 77008-920
6-Bromo-N,N-dimethylpyridin-3-amine 95%
6-Bromo-N,N-dimethylpyridin-3-amine
Precaución: for Research Use Only


Formula: C₇H₉BrN₂
MDL Number: MFCD00100094
CAS Number: 39856-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty