Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-9,9-diphenylfluorene 98%

Supplier: AMBEED, INC
Warning

A112312-100G A112312-10G A112312-1G A112312-25G A112312-5G
77025-828EA 127 USD InStock
77025-828 77025-830 77025-832 77025-834 77025-836 77025-838
2-Bromo-9,9-diphenylfluorene 98%
2-Bromo-9,9-diphenylfluorene
Precaución: for Research Use Only


Formula: C₂₅H₁₇Br
MDL Number: MFCD09907957
CAS Number: 474918-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty