Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5-difluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A112627-100MG A112627-1G A112627-250MG
77008-988EA 41 USD InStock
77008-988 77008-990 77008-992
3-Bromo-2,5-difluorobenzonitrile 98%
3-Bromo-2,5-difluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₂BrF₂N
MDL Number: MFCD27923485
CAS Number: 1638487-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty