Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,5-dimethyl-4-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A112879-10G A112879-1G A112879-250MG A112879-25G A112879-5G
77009-010EA 51 USD InStock
77009-010 77009-012 77009-014 77009-016 77009-018
1-Bromo-2,5-dimethyl-4-nitrobenzene 98%
1-Bromo-2,5-dimethyl-4-nitrobenzene
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
MDL Number: MFCD01463298
CAS Number: 15540-81-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty