Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4,6-dichloro-5-nitronicotinate 97%

Supplier: AMBEED, INC
Warning

A112987-100MG A112987-250MG
76995-326EA 13 USD InStock
76995-326 76995-328
Ethyl-4,6-dichloro-5-nitronicotinate 97%
Ethyl-4,6-dichloro-5-nitronicotinate
Precaución: for Research Use Only


Formula: C₈H₆Cl₂N₂O₄
MW: 265.05 g/mol
MDL Number: MFCD09842620
CAS Number: 154012-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty