Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5'-Chloro-2'-hydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A113905-100G A113905-10G A113905-25G A113905-500G A113905-5G
76995-390EA 77 USD InStock
76995-390 76995-392 76995-394 76995-396 76995-398
5'-Chloro-2'-hydroxyacetophenone 98%
5'-Chloro-2'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MDL Number: MFCD00067788
CAS Number: 1450-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty