Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tyrphostin AG 1024 98%

Supplier: AMBEED, INC
Warning

A114134-100MG A114134-10MG A114134-1MG A114134-25MG A114134-50MG A114134-5MG
77009-084EA 314 USD InStock
77009-084 77009-086 77009-088 77009-090 77009-092 77009-094
Tyrphostin AG 1024 98%
Tyrphostin AG 1024
Precaución: for Research Use Only


Formula: C₁₄H₁₃BrN₂O
MW: 305.2 g/mol
MDL Number: MFCD02179365
CAS Number: 65678-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty