Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3,4-Dihydroxy-2-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A114883-100G A114883-1G A114883-250MG A114883-5G
77009-124EA 479 USD InStock
77009-124 77009-126 77009-128 77009-130
Methyl 3,4-Dihydroxy-2-methylbenzoate 98%
Methyl 3,4-Dihydroxy-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD16250128
CAS Number: 740799-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty