Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A114930-10G A114930-1G A114930-5G
76982-916EA 166 USD InStock
76982-916 76982-918 76982-920
4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester 97%
4-(1-Pyrrolidinylmethyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₇H₂₆BNO₂
MDL Number: MFCD08690299
CAS Number: 884507-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty