Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid 98%

Supplier: AMBEED, INC
Warning

A115166-10G A115166-1G A115166-5G
77026-040EA 85 USD InStock
77026-040 77026-042 77026-044
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid 98%
2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₄
MDL Number: MFCD13152279
CAS Number: 178306-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty