Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-thenoic acid 97%

Supplier: AMBEED, INC
Warning

A116048-100G A116048-10G A116048-1G A116048-25G A116048-500G A116048-5G
76982-966EA 72 USD InStock
76982-966 76982-968 76982-970 76982-972 76982-974 76982-976
3-Bromo-2-thenoic acid 97%
3-Bromo-2-thenoic acid
Precaución: for Research Use Only


Formula: C₅H₃BrO₂S
MDL Number: MFCD00052291
CAS Number: 7311-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty