Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromo-5-(trifluoromethyl)phenyl)methanol 97%

Supplier: AMBEED, INC
Warning

A116593-1G A116593-250MG A116593-25G A116593-5G
77026-142EA 8 USD InStock
77026-142 77026-144 77026-146 77026-148
(3-Bromo-5-(trifluoromethyl)phenyl)methanol 97%
(3-Bromo-5-(trifluoromethyl)phenyl)methanol
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O
MW: 255.0300 g/mol
MDL Number: MFCD09835207
CAS Number: 172023-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty