Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A-769662 ≥98%

Supplier: AMBEED, INC
Warning

A117039-10MG A117039-1MG A117039-50MG A117039-5MG
77026-172EA 31 USD InStock
77026-172 77026-174 77026-176 77026-178
A-769662 ≥98%
A-769662
Precaución: for Research Use Only


Formula: C₂₀H₁₂N₂O₃S
MDL Number: MFCD11977269
CAS Number: 844499-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty