Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Bromophenyl)piperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A117792-10G A117792-1G A117792-250MG
77026-220EA 169 USD InStock
77026-220 77026-222 77026-224
4-(4-Bromophenyl)piperidine hydrochloride 95%
4-(4-Bromophenyl)piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₅BrClN
MDL Number: MFCD08461756
CAS Number: 769944-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty