Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A118703-1G A118703-250MG A118703-25G A118703-5G
77026-262EA 6 USD InStock
77026-262 77026-264 77026-266 77026-268
5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde 97%
5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde
Precaución: for Research Use Only


Formula: C₂₆H₁₇ClFN₃O₃
MW: 473.89 g/mol
MDL Number: MFCD10565688
CAS Number: 231278-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty