Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5-dichloropyridine 95%

Supplier: AMBEED, INC
Warning

A118770-100G A118770-10G A118770-1G A118770-25G A118770-5G
76995-664EA 167 USD InStock
76995-664 76995-666 76995-668 76995-670 76995-672
3-Bromo-2,5-dichloropyridine 95%
3-Bromo-2,5-dichloropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrCl₂N
MW: 226.89
MDL Number: MFCD06659484
CAS Number: 138006-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty