Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KU-0063794 ≥98%

Supplier: AMBEED, INC
Warning

A119369-10MG A119369-1MG A119369-5MG
77026-282EA 69 USD InStock
77026-282 77026-284 77026-286
KU-0063794 ≥98%
KU-0063794
Precaución: for Research Use Only


Formula: C₂₅H₃₁N₅O₄
MW: 465.6 g/mol
MDL Number: MFCD22666594
CAS Number: 938440-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty