Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethyl)anisole 98+%

Supplier: AMBEED, INC
Warning

A119612-1G A119612-25G A119612-5G
76983-166EA 11 USD InStock
76983-166 76983-168 76983-170
2-Bromo-4-(trifluoromethyl)anisole 98+%
2-Bromo-4-(trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O
MW: 255.03
MDL Number: MFCD03789249
CAS Number: 402-10-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty