Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Iodobenzo[d]thiazole 97%

Supplier: AMBEED, INC
Warning

A119690-1G A119690-100MG A119690-250MG
77026-310EA 143 USD InStock
77026-310 77666-714 77666-716
6-Iodobenzo[d]thiazole 97%
6-Iodobenzo[d]thiazole
Precaución: for Research Use Only


Formula: C₇H₄INS
MW: 261.08 g/mol
MDL Number: MFCD06659655
CAS Number: 654070-00-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty