Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chlorobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A120655-100G A120655-10G A120655-1G A120655-25G A120655-5G
77009-416EA 55 USD InStock
77009-416 77009-418 77009-420 77009-422 77009-424
5-Bromo-2-chlorobenzaldehyde 98%
5-Bromo-2-chlorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD08445659
CAS Number: 189628-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty