Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzothiophene-6-carboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A120837-100MG A120837-1G A120837-250MG A120837-50MG
76983-252EA 35 USD InStock
76983-252 76983-254 76983-256 76983-258
1-Benzothiophene-6-carboxaldehyde 98%
1-Benzothiophene-6-carboxaldehyde
Precaución: for Research Use Only


Formula: C₉H₆OS
MW: 162.208 g/mol
MDL Number: MFCD18451824
CAS Number: 6386-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty