Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxycinnamic acid 98%

Supplier: AMBEED, INC
Warning

A121736-1000G A121736-100G A121736-10G A121736-25G A121736-500G
77026-430EA 144 USD InStock
77026-430 77026-432 77026-434 77026-436 77026-438 77026-440
4-Methoxycinnamic acid 98%
4-Methoxycinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MDL Number: MFCD00004398
CAS Number: 830-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty