Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Isoleucine methyl ester hydrochloride 98%

Supplier: AMBEED, INC
Warning

A121861-100G A121861-10G A121861-25G A121861-5G A121861-500G
76995-846EA 46 USD InStock
76995-846 76995-848 76995-850 76995-852 77670-724
L-Isoleucine methyl ester hydrochloride 98%
L-Isoleucine methyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₅NO₂
MDL Number: MFCD00038911
CAS Number: 18598-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty