Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazolo[5,4-d]pyrimidin-7-ol 95%

Supplier: AMBEED, INC
Warning

A122212-100MG A122212-250MG
77009-466EA 168 USD InStock
77009-466 77009-468
Thiazolo[5,4-d]pyrimidin-7-ol 95%
Thiazolo[5,4-d]pyrimidin-7-ol
Precaución: for Research Use Only


Formula: C₅H₃N₃OS
MDL Number: MFCD13195519
CAS Number: 5021-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty