Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,3-Tetrafluoro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A122269-1G A122269-25G A122269-5G
77026-484EA 15 USD InStock
77026-484 77026-486 77026-488
α,α,α,3-Tetrafluoro-o-toluic acid 98%
α,α,α,3-Tetrafluoro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₄F₄O₂
MW: 208.11
MDL Number: MFCD01631565
CAS Number: 261951-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty