Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate 95%

Supplier: AMBEED, INC
Warning

A122412-10G A122412-1G A122412-25G A122412-5G
77026-498EA 36 USD InStock
77026-498 77026-500 77026-502 77026-504
(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate 95%
(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate
Precaución: for Research Use Only


Formula: C₂₃H₃₇N₃O₅
MW: 435.57 g/mol
MDL Number: MFCD09840998
CAS Number: 219921-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty