Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Bromomethyl)phenyl)pyridine 95%

Supplier: AMBEED, INC
Danger

A122898-100MG A122898-250MG
76995-936EA 108 USD InStock
76995-936 76995-938
2-(4-(Bromomethyl)phenyl)pyridine 95%
2-(4-(Bromomethyl)phenyl)pyridine
Precaución: for Research Use Only


Formula: C₁₂H₁₀BrN
MDL Number: MFCD06659494
CAS Number: 52199-24-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty