Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A123176-10G A123176-1G A123176-250MG A123176-5G
76995-942EA 40 USD InStock
76995-942 76995-944 76995-946 76995-948
2-Chloro-4-methoxyphenylboronic acid 98%
2-Chloro-4-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₃
MDL Number: MFCD03411935
CAS Number: 219735-99-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty