Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4'-Pentyl-(1,1'-bicyclohexyl)-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A123557-100G A123557-1G A123557-25G A123557-5G
77026-622EA 97 USD InStock
77026-622 77026-624 77026-626 77026-628
trans-4'-Pentyl-(1,1'-bicyclohexyl)-4-carboxylic acid 97%
trans-4'-Pentyl-(1,1'-bicyclohexyl)-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₈H₃₂O₂
MDL Number: MFCD22379991
CAS Number: 65355-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty