Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Bromopyridin-3-yl)pivalamide 95+%

Supplier: AMBEED, INC
Warning

A124082-100MG A124082-1G A124082-250MG A124082-5G
76995-970EA 5 USD InStock
76995-970 76995-972 76995-974 76995-976
N-(2-Bromopyridin-3-yl)pivalamide 95+%
N-(2-Bromopyridin-3-yl)pivalamide
Precaución: for Research Use Only


Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
MDL Number: MFCD07367912
CAS Number: 835882-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty