Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A124509-250MG A124509-1G
77009-580EA 31 USD InStock
77009-580 77683-110
tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate 98%, Ambeed.Inc
tert-Butyl-4-(cyanomethylidene)piperidine-1-carboxylate
Precaución: For research use only.


Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD03695496
CAS Number: 197569-11-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty