Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzoyl-2'-deoxyadenosine 98%

Supplier: AMBEED, INC
Warning

A124703-100G A124703-10G A124703-1G A124703-25G A124703-5G
76996-032EA 563 USD InStock
76996-032 76996-034 76996-036 76996-038 76996-040
N-Benzoyl-2'-deoxyadenosine 98%
N-Benzoyl-2'-deoxyadenosine
Precaución: for Research Use Only


Formula: C₁₇H₁₇N₅O₄
MDL Number: MFCD00009628
CAS Number: 4546-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty