Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-2-hydroxypyridine 97%

Supplier: AMBEED, INC
Warning

A125856-100G A125856-10G A125856-1G A125856-25G A125856-5G
76996-094EA 72 USD InStock
76996-094 76996-096 76996-098 76996-100 76996-102
3-Cyano-2-hydroxypyridine 97%
3-Cyano-2-hydroxypyridine
Precaución: for Research Use Only


Formula: C6H4N2O
MDL Number: MFCD00661865
CAS Number: 20577-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty