Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Trifluoromethoxy)indoline 98%

Supplier: AMBEED, INC
Warning

A126284-100MG A126284-1G A126284-250MG
77009-660EA 15 USD InStock
77009-660 77009-662 77009-664
6-(Trifluoromethoxy)indoline 98%
6-(Trifluoromethoxy)indoline
Precaución: for Research Use Only


Formula: C₉H₈F₃NO
MDL Number: MFCD09954774
CAS Number: 959235-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty