Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-3-hydroxypropanoate 95%

Supplier: AMBEED, INC
Warning

A126883-100MG A126883-1G A126883-250MG A126883-5G
77009-704EA 44 USD InStock
77009-704 77009-706 77009-708 77009-710
Methyl 2-bromo-3-hydroxypropanoate 95%
Methyl 2-bromo-3-hydroxypropanoate
Precaución: for Research Use Only


Formula: C₄H₇BrO₃
MW: 183.00 g/mol
MDL Number: MFCD20486781
CAS Number: 7691-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty