Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-1-benzofuran 96%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A127583-100MG A127583-1G A127583-250MG A127583-5G
76983-518EA 66 USD InStock
76983-518 76983-520 76983-522 77683-130
5-Nitro-1-benzofuran 96%, Ambeed.Inc
5-Nitro-1-benzofuran
Precaución: For research use only.


Formula: C₈H₅NO₃
MDL Number: MFCD00456542
CAS Number: 18761-31-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty