Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorophenyl)acrylaldehyde 95% (by GC)

Supplier: AMBEED, INC
Warning

A127670-25G A127670-5G
76996-168EA 60 USD InStock
76996-168 76996-170
3-(4-Fluorophenyl)acrylaldehyde 95% (by GC)
3-(4-Fluorophenyl)acrylaldehyde
Precaución: for Research Use Only


Formula: C₉H₇FO
MDL Number: MFCD03425895
CAS Number: 24654-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty