Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mitiglinide calcium 98%

Supplier: AMBEED, INC
Warning

A127678-100MG A127678-250MG A127678-25MG
76996-172EA 8 USD InStock
76996-172 76996-174 76996-176
Mitiglinide calcium 98%
Mitiglinide calcium
Precaución: for Research Use Only


Formula: C₃₈H₄₈CaN₂O₆
MW: 668.87 g/mol
MDL Number: MFCD09263183
CAS Number: 145525-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty