Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(1-Boc-piperidino)-4-iodo-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A127688-1G A127688-25G A127688-5G A127688-250MG
77026-904EA 6 USD InStock
77026-904 77026-906 77026-908 77666-820
1-(1-Boc-piperidino)-4-iodo-1H-pyrazole 97%
1-(1-Boc-piperidino)-4-iodo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₃H₂₀IN₃O₂
MDL Number: MFCD12913927
CAS Number: 877399-73-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty