Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-nitrophenyl)amine 97%

Supplier: AMBEED, INC
Warning

A128635-100G A128635-10G A128635-1G A128635-250MG A128635-25G A128635-5G
76996-236EA 133 USD InStock
76996-236 76996-238 76996-240 76996-242 76996-244 76996-246
Tris(4-nitrophenyl)amine 97%
Tris(4-nitrophenyl)amine
Precaución: for Research Use Only


Formula: C₁₈H₁₂N₄O₆
MDL Number: MFCD00024636
CAS Number: 20440-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty