Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-iodobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A129861-10G A129861-250MG A129861-25G A129861-5G
76983-622EA 95 USD InStock
76983-622 76983-624 76983-626 76983-628
4-Chloro-2-iodobenzonitrile 98%
4-Chloro-2-iodobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClIN
MDL Number: MFCD09025673
CAS Number: 61272-75-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty