Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-tert-Butylbenzonitrile 97%

Supplier: AMBEED, INC
Warning

A129876-100G A129876-10G A129876-1G A129876-25G A129876-5G
76983-630EA 285 USD InStock
76983-630 76983-632 76983-634 76983-636 76983-638
3-tert-Butylbenzonitrile 97%
3-tert-Butylbenzonitrile
Precaución: for Research Use Only


Formula: C₁₁H₁₃N
MW: 159.23 g/mol
MDL Number: MFCD08448298
CAS Number: 154532-34-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty