Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Pyrazol-1-ylphenyl)methanol 98%

Supplier: AMBEED, INC
Warning

A130006-10G A130006-1G A130006-25G
76996-326EA 41 USD InStock
76996-326 76996-328 76996-330
(4-Pyrazol-1-ylphenyl)methanol 98%
(4-Pyrazol-1-ylphenyl)methanol
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
MDL Number: MFCD02682057
CAS Number: 143426-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty