Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetylsalicylsalicylic acid 95%

Supplier: AMBEED, INC
Warning

A130448-100MG A130448-1G A130448-250MG A130448-5G
77027-036EA 29 USD InStock
77027-036 77027-038 77027-040 77027-042
Acetylsalicylsalicylic acid 95%
Acetylsalicylsalicylic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₂O₆
MDL Number: MFCD00143537
CAS Number: 530-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty